Methyl-2,5-dihydroxybenzoate, Sigma-Aldrich®
Catalog # SIAL426091-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:168.15 g/mol
EINECS:218-427-8
CAS Number:2150-46-1
Formula:C₈H₈O₄
MDL Number:MFCD00016464