Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tris(2,4,6-trimethoxyphenyl)phosphine
Tris(2,4,6-trimethoxyphenyl)phosphine, Sigma-Aldrich®
Tris(2,4,6-trimethoxyphenyl)phosphine, Sigma-Aldrich®
Tris(2,4,6-trimethoxyphenyl)phosphine, Sigma-Aldrich®
SIAL392081-5G
:
Merck
:
Tris(2,4,6-trimethoxyphenyl)phosphine, Sigma-Aldrich®
SIAL392081-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
532.519 g/mol
:
91608-15-0
:
C₂₇H₃₃O₉P
:
MFCD00145467