Sodium tetrachloropalladate(II) ≥99.995% (metals basis), Sigma-Aldrich®
Catalog # SIAL379808-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:294.21 g/mol
:M-Clarity™ quality level = MQ100
EINECS:237-502-6
CAS Number:13820-53-6
Formula:Cl₄Na₂Pd
MDL Number:MFCD00003487