Oleyl alcohol, Sigma-Aldrich®
SIAL369314-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:268.48 g/mol
:12,06968
:333 °C (1013 hPa)
:13…19 °C
:0.85 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:205-597-3
:143-28-2
:C₁₈H₃₆O
:MFCD00002993