Tris(trimethylsilyl)silane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1873-77-4
:
Formula:C₉H₂₈Si₄
MDL Number:MFCD00077893
:248.66 g/mol
Merck Index:12,09886
Boiling Pt:82 °C (1013 hPa)
Density:0.8 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100