Order Entry
Export
ContactUsLinkComponent
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, Sigma-Aldrich®
Irritant
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, Sigma-Aldrich®
Catalog # SIAL32869-50G
Supplier:  Merck
CAS Number:  
Irritant
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, Sigma-Aldrich®
Catalog # SIAL32869-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :364.35 g/mol
Melting Pt:213…216 °C
EINECS:236-308-9
 :GHS07
Storage Temperature:Ambient
CAS Number:125572-95-4
 :MFCD00066429
Formula:C₁₄H₂₂N₂O₈·1H₂O