
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:125572-95-4
:MFCD00066429
Formula:C₁₄H₂₂N₂O₈·1H₂O
:364.35 g/mol
Melting Pt:213…216 °C
:M-Clarity™ quality level = MQ200
EINECS:236-308-9
:GHS07
Storage Temperature:Ambient