Order Entry
Export
ContactUsLinkComponent
2',4'-Dihydroxy-3'-methylacetophenone 90%, Technical Grade, Sigma-Aldrich®
Irritant
2',4'-Dihydroxy-3'-methylacetophenone 90%, Technical Grade, Sigma-Aldrich®
Catalog # 296244-5G
CAS Number:  
undefined
Irritant
2',4'-Dihydroxy-3'-methylacetophenone 90%, Technical Grade, Sigma-Aldrich®
Catalog # 296244-5G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Yellow to orange to brown powder or crystals
  • Infrared spectrum:
    Conforms to structure
  • Gas liquid:
    88.0% (minimum)
    Approved September 12, 2008 RJM

Specification Test Results

About this item

 :
Melting Pt:153…156 °C (lit.)
 :M-Clarity™ quality level = MQ100
 :166.18 g/mol
 :GHS07
Storage Temperature:Room Temperature
CAS Number:10139-84-1
Formula:C₉H₁₀O₃
MDL Number:MFCD00010817