α,α,α,α',α',α'-Hexafluoro-p-xylene, Sigma-Aldrich®
Catalog # SIAL290912-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:214.11 g/mol
Boiling Pt:113…115 °C (1013 hPa)
Melting Pt:3 °C
Density:1.3808 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:207-086-0
CAS Number:433-19-2
Formula:C₈H₄F₆
MDL Number:MFCD00000402