Order Entry
Export
ContactUsLinkComponent
1,1-Dimethylurea, Sigma-Aldrich®
1,1-Dimethylurea, Sigma-Aldrich®
Catalog # SIAL261394-25G
Supplier:  Merck
CAS Number:  
1,1-Dimethylurea, Sigma-Aldrich®
Catalog # SIAL261394-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :88.11 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:209-957-0
CAS Number:598-94-7
Formula:C₃H₈N₂O
MDL Number:MFCD00007959