Order Entry
Export
Orders LinkContactUsLinkComponent
Nitric acid 70% ≥70%

Nitric acid 70% ≥70%

6×500 g, 1
Catalog # SIAL258113-6X500G
Oxidizing
Corrosive
Supplier:   Merck
Nitric acid 70% ≥70%
Catalog # SIAL258113-6X500G
Supplier:  Merck
CAS Number:  7697-37-2
Nitric acid 70% ≥70%

6×500 g, 1

CAS Number:  7697-37-2
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents
Synonyms:  Nitric acid solution
  • CAS Number:7697-37-2
  • Formula:HNO₃
  • MDL Number:MFCD00011349
  • Molecular Weight:63.01 g/mol
  • Storage Temperature:Ambient
  • ADR:8,I
  •  :M-Clarity™ quality level = MQ200
  •  :Density: 1.51 g/cm³ (20 °C)
  • EINECS:231-714-2
  • UN:2031
Specifications

Specifications

  • Pk:
    1
  • Pk Info:
    6×500 g

Specifications