Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzyltrimethylammonium chloride
Benzyltrimethylammonium chloride, Sigma-Aldrich®
Benzyltrimethylammonium chloride, Sigma-Aldrich®
:
Merck
Benzyltrimethylammonium chloride, Sigma-Aldrich®
:
Merck
:
56-93-9
:
:
C₁₀H₁₆ClN
:
MFCD00011782
:
185.7 g/mol
:
238 °C
:
1.07 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ200
:
200-300-3
:
GHS07
:
Ambient