Order Entry
Export
ContactUsLinkComponent
Nitric acid ≥70%, Sigma-Aldrich®
Oxidizing
Corrosive
Nitric acid ≥70%, Sigma-Aldrich®
Catalog # SIAL225711-475ML
Supplier:  Merck
CAS Number:  
Oxidizing
Corrosive
Nitric acid ≥70%, Sigma-Aldrich®
Catalog # SIAL225711-475ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    475 mL
  • Seq:
    2710

Specifications

About this item

 :
 :63.01 g/mol
ADR:8,I
 :M-Clarity™ quality level = MQ100
EINECS:231-714-2
UN:2031
 :GHS03
 :GHS05
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349