4,4'-Difluorobenzhydrol, Sigma-Aldrich®
Catalog # SIAL222682-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:220.22 g/mol
Boiling Pt:305 °C (1013 hPa)
Melting Pt:47 °C
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:206-671-8
CAS Number:365-24-2
Formula:C₁₃H₁₀F₂O
MDL Number:MFCD00000357