Order Entry
Export
ContactUsLinkComponent
Phenosafranine 80% (dye content), Sigma-Aldrich®
Irritant
Phenosafranine 80% (dye content), Sigma-Aldrich®
Catalog # 199648-1G
CAS Number:  
undefined
Irritant
Phenosafranine 80% (dye content), Sigma-Aldrich®
Catalog # 199648-1G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    ≥46.7%
  • Nitrogen:
    ≥12.0%
  • Wavelength:
    270 - 276 nm
  • Extinction Coefficient:
    ≥30000

Specification Test Results

About this item

 :
 :322.8 g/mol
Merck Index:12,07402
Melting Pt:>300 °C
EINECS:201-387-0
 :GHS07
Storage Temperature:Room Temperature
CAS Number:81-93-6
Formula:C₁₈H₁₅ClN₄