2-(4-Methylthiazol-5-yl)ethanol, Sigma-Aldrich®
Catalog # SIAL190675-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:143.21 g/mol
Merck Index:12,06208
Boiling Pt:135 °C (1013 hPa)
Density:1.199 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:205-272-6
CAS Number:137-00-8
Formula:C₆H₉NOS
MDL Number:MFCD00005339