Methyl propiolate, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
- CAS Number:922-67-8
- Formula:C₄H₄O₂
- MDL Number:MFCD00008572
- Molecular Weight:84.07 g/mol
- Boiling Pt:105 °C (1013 hPa)
- Density:1.196 g/cm³ (20 °C)
- EINECS:213-083-5
- UN:3272
- ADR:3,II
- :M-Clarity™ quality level = MQ200
3 Options Available Below
Product Details & Documents
Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
- CAS Number:922-67-8
- Formula:C₄H₄O₂
- MDL Number:MFCD00008572
- Molecular Weight:84.07 g/mol
- Boiling Pt:105 °C (1013 hPa)
- Density:1.196 g/cm³ (20 °C)
- EINECS:213-083-5
- UN:3272
- ADR:3,II
- :M-Clarity™ quality level = MQ200



