α'-Chloro-α,α,α-trifluoro-m-xylene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:705-29-3
:
Formula:C₈H₆ClF₃
MDL Number:MFCD00000908
:194.58 g/mol
Boiling Pt:95 °C (1013 hPa)
Density:1.3079 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-884-4
UN:3265
ADR:8,III
:M-Clarity™ quality level = MQ100