4,4'-Dibromobenzophenone, Sigma-Aldrich®
Catalog # SIAL166634-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:340.01 g/mol
:M-Clarity™ quality level = MQ100
EINECS:223-632-0
CAS Number:3988-03-2
Formula:C₁₃H₈Br₂O
MDL Number:MFCD00016330