Undec-10-enoyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:38460-95-6
:
Formula:C₁₁H₁₉ClO
MDL Number:MFCD00000772
:202.72 g/mol
Boiling Pt:120 °C (1013 hPa)
Density:0.94 g/cm³ (20 °C)
EINECS:253-951-0
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS07