4,6-Dichloropyrimidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1193-21-1
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00006109
:148.98 g/mol
Boiling Pt:176 °C (1013 hPa)
Melting Pt:64…66 °C
Density:1.734 g/cm³ (20 °C)
EINECS:214-770-2
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:3261