1-Ethyl-3-methylimidazoliumtetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:143314-16-3
:
Formula:C₆H₁₁BF₄N₂
MDL Number:MFCD00216668
:197.97 g/mol
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:∼15 °C
Density:1.34 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:>110 °C