Order Entry
Export
Orders LinkContactUsLinkComponent
1-Octadecanol

1-Octadecanol

Glass bottle for solids, 1 kg
Catalog # ROTH8663.1
Supplier:   Roth Carl
1-Octadecanol
Catalog # ROTH8663.1
Supplier:  Roth Carl
CAS Number:  112-92-5
1-Octadecanol

Glass bottle for solids, 1 kg

CAS Number:  112-92-5
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
  • CAS Number:112-92-5
  • Formula:H₃C(CH₂)₁₆CH₂OH
  • MDL Number:MFCD00002823
  • Molecular Weight:270.5 g/mol
  • Merck Index:12,08960
  • Boiling Pt:336 °C (1013 hPa)
  • Melting Pt:56…57 °C
  • Density:0.805…0.815 g/cm³ (60 °C)
  • Storage Temperature:Ambient
  • EINECS:204-017-6

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications