Indol-3-acetic acid
Catalog # ROTH7280.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:175.19 g/mol
Merck Index:13,04986
Boiling Pt:120…125 °C (0.05 mmHg)
Melting Pt:167…170 °C
Storage Temperature:Refrigerator
EINECS:201-748-2
CAS Number:87-51-4
Formula:C₁₀H₉NO₂
MDL Number:MFCD00005636