Order Entry
Export
Orders LinkContactUsLinkComponent
4-Hydroxybenzaldehyde

4-Hydroxybenzaldehyde

Glass bottle for solids, 250 g
Catalog # ROTH3870.1
Irritant
Supplier:   Roth Carl
4-Hydroxybenzaldehyde
Catalog # ROTH3870.1
Supplier:  Roth Carl
CAS Number:  123-08-0
4-Hydroxybenzaldehyde

Glass bottle for solids, 250 g

CAS Number:  123-08-0
Price
104,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:123-08-0
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00006939
  • Molecular Weight:122.12 g/mol
  • Merck Index:13,04836
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:116 °C
  • Density:1.13 g/cm³ (20 °C)
  • Flash Pt:174 °C
  • Storage Temperature:Ambient
  • EINECS:204-599-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications