Order Entry
Export
ContactUsLinkComponent
di-Sodium succinate hexahydrate
di-Sodium succinate hexahydrate
Catalog # ROTH3195.1
Supplier:  Roth Carl
CAS Number:  
di-Sodium succinate hexahydrate
Catalog # ROTH3195.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :270.14 g/mol
Merck Index:13,08754
Melting Pt:300 °C
Density:1.529 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-778-7
CAS Number:6106-21-4
Formula:NaOOCCH₂CH₂COONa·6H₂O
MDL Number:MFCD00149117