Squalene
Glass bottle for solids, 1 g
Catalog # ROTH3185.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- MDL Number:MFCD00008912
- Molecular Weight:410.73 g/mol
- Merck Index:13,08847
- Boiling Pt:429 °C (1013 hPa)
- Melting Pt:–5 °C
- Density:0.86 g/cm³ (20 °C)
- Flash Pt:200 °C
- EINECS:203-826-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Cat. No.:ROTH3185.1



