2,3-Diaminophenazine
Glass bottle for solids, 100 mg
Catalog # PROOMM2500.01
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: DAP, Phenazine-2,3-diyldiamine
- CAS Number:655-86-7
- Formula:C₁₂H₁₀N₄
- MDL Number:MFCD00160693
- Molecular Weight:210.24 g/mol
- EINECS:211-512-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



