ω-Chloro-2,4-difluoroacetophenone
Catalog # PROOMM0235.04
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:190.58 g/mol
Boiling Pt:233 °C (1013 hPa)
Melting Pt:47…48 °C
UN:3261
ADR:8,II
CAS Number:51336-94-8
Formula:C₈H₅ClF₂O
MDL Number:MFCD00013252