Order Entry
Export
ContactUsLinkComponent
(3,4-Dimethoxyphenyl)acetonitrile
(3,4-Dimethoxyphenyl)acetonitrile
Catalog # PROOMM0022.18
CAS Number:  
(3,4-Dimethoxyphenyl)acetonitrile
Catalog # PROOMM0022.18
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :177.2
Boiling Pt:320 °C (1013 hPa)
Melting Pt:60…62 °C
EINECS:202-225-1
CAS Number:93-17-4
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD00001911