2-(4-Methylthiazol-5-yl)ethanol
Catalog # PROOBP444
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:143.21 g/mol
Merck Index:12,06208
Boiling Pt:135 °C (1013 hPa)
Density:1.199 g/cm³ (20 °C)
EINECS:205-272-6
CAS Number:137-00-8
Formula:C₆H₉NOS
MDL Number:MFCD00005339