
1,3-Dihydroxynaphthalene ≥99%
Catalog # 25771.126
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Assay:Min. 99 %
Specification Test Results
About this item
:
:160.17 g/mol
Merck Index:12,06483
Boiling Pt:386 °C (1013 hPa)
Melting Pt:124 °C
Density:1.12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-079-7
:GHS07
CAS Number:132-86-5
Formula:C₁₀H₈O₂
MDL Number:MFCD00003965