Order Entry
Export
Orders LinkContactUsLinkComponent
5-Ethyluracil

5-Ethyluracil

Glass bottle for solids, 1 g
Catalog # MOLE38127331-1G
Supplier:   Molekula
5-Ethyluracil
Catalog # MOLE38127331-1G
Supplier:  Molekula
CAS Number:  4212-49-1
5-Ethyluracil

Glass bottle for solids, 1 g

CAS Number:  4212-49-1
Price
73,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Ethyluracil, 5-Ethylpyrimidine-2,4(1H,3H)-dione
  • CAS Number:4212-49-1
  • Formula:C₆H₈N₂O₂
  • MDL Number:MFCD00079187
  • Molecular Weight:140.14 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications