Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Chlorobenzhydrol

Glass bottle for solids, 25 g
Catalog # MOLE25057386-25G
Supplier:   Molekula
4-Chlorobenzhydrol
Catalog # MOLE25057386-25G
Supplier:  Molekula
CAS Number:  119-56-2
4-Chlorobenzhydrol

Glass bottle for solids, 25 g

CAS Number:  119-56-2
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4-Chlorophenyl phenyl carbinol, 4-Chlorobenzhydryl alcohol, 4-Chloro-α-phenylbenzyl alcohol, p-Chlorobenzhydrol, 4-Chlorodiphenylmethanol
  • CAS Number:119-56-2
  • Formula:C₁₃H₁₁ClO
  • MDL Number:MFCD00004491
  • Molecular Weight:218.68 g/mol
  • EINECS:204-333-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications