Order Entry
Export
ContactUsLinkComponent
Tris(diethylamino)phosphine
Tris(diethylamino)phosphine
Catalog # MOLE60554819-25G
Supplier:  Molekula
CAS Number:  
Tris(diethylamino)phosphine
Catalog # MOLE60554819-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :247.36 g/mol
Density:0.89 g/cm³ (20 °C)
EINECS:218-920-8
CAS Number:2283-11-6
Formula:C₁₂H₃₀N₃P
MDL Number:MFCD00009041