Order Entry
Export
ContactUsLinkComponent
 
1,4-Dimethylpiperazine
Catalog # MOLE15044534-100G
Supplier:  Molekula
CAS Number:  
1,4-Dimethylpiperazine
Catalog # MOLE15044534-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :114.19 g/mol
Boiling Pt:131 °C (1013 hPa)
Melting Pt:–1 °C
Density:0,85 g/cm³ (20 °C)
EINECS:203-412-0
CAS Number:106-58-1
Formula:C₆H₁₄N₂
MDL Number:MFCD00006156