4,6-Dichloropyrimidine
Catalog # MOLEM61506519
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:148.98 g/mol
Boiling Pt:176 °C (1013 hPa)
Melting Pt:64…66 °C
Density:1.734 g/cm³ (20 °C)
EINECS:214-770-2
UN:3261
ADR:8,III
CAS Number:1193-21-1
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00006109