
4,4'-Difluorobenzophenone
Catalog # MOLE23433514-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Boiling Pt:170…172 °C (10 mmHg)
Melting Pt:140…141 °C
Density:1,39 g/cm³ (20 °C)
:218.2 g/mol
EINECS:206-466-3
:GHS07
CAS Number:345-92-6
Formula:C₁₃H₈F₂O
MDL Number:MFCD00000353