Order Entry
Export
ContactUsLinkComponent
2-Aminothiazole
2-Aminothiazole
Catalog # MOLE10637809-500G
Supplier:  Molekula
CAS Number:  
2-Aminothiazole
Catalog # MOLE10637809-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Thiazol-2-ylamine, Aminothiazole, Thiazol-2-amine
 :
 :100.14 g/mol
Merck Index:13,00479
Boiling Pt:216 °C (1013 hPa)
Melting Pt:86…89 °C
Density:1.5 g/cm³ (20 °C)
EINECS:202-511-6
CAS Number:96-50-4
Formula:C₃H₄N₂S
MDL Number:MFCD00005325