2,4-Dimethoxypyrimidine
Catalog # MOLE58183615-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:140.14 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:17.5 °C
CAS Number:3551-55-1
Formula:C₆H₈N₂O₂
MDL Number:MFCD00056028
Boiling Pt:205 °C (1013 hPa)
Melting Pt:17,5 °C