(3,4-Dimethoxyphenyl)acetonitrile
Catalog # MOLEM47923583
Supplier: Molekula
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:177.2
Boiling Pt:320 °C (1013 hPa)
Melting Pt:60…62 °C
EINECS:202-225-1
CAS Number:93-17-4
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD00001911