Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl propiolate

Ethyl propiolate

Glass bottle for solids, 25 g
Catalog # MOLEM41170359
Supplier:   Molekula
Ethyl propiolate
Catalog # MOLEM41170359
Supplier:  Molekula
CAS Number:  623-47-2
Ethyl propiolate

Glass bottle for solids, 25 g

CAS Number:  623-47-2
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester
  • CAS Number:623-47-2
  • Formula:C₅H₆O₂
  • MDL Number:MFCD00009184
  • Molecular Weight:98.1 g/mol
  • Boiling Pt:120 °C (1013 hPa)
  • Melting Pt:9 °C
  • Density:0.9788 g/cm³ (4 °C)
  • EINECS:210-795-8
  • UN:3272
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications