Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl propiolate

Methyl propiolate

Glass bottle for solids, 25 g
Catalog # MOLE14690909-25G
Supplier:   Molekula
Methyl propiolate
Catalog # MOLE14690909-25G
Supplier:  Molekula
CAS Number:  922-67-8
Methyl propiolate

Glass bottle for solids, 25 g

CAS Number:  922-67-8
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
  • CAS Number:922-67-8
  • Formula:C₄H₄O₂
  • MDL Number:MFCD00008572
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:105 °C (1013 hPa)
  • Density:1.196 g/cm³ (20 °C)
  • EINECS:213-083-5
  • UN:3272
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications