5,6-Dimethylbenzimidazole
Glass bottle for solids, 100 g
Catalog # MOLEM35343197
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 5,6-Dimethyl-1H-benzimidazole, 5,6-Dimethyl-1H-benzo[d]imidazole
- CAS Number:582-60-5
- Formula:C₉H₁₀N₂
- MDL Number:MFCD00005603
- Molecular Weight:146.19 g/mol
- Merck Index:13,03263
- Boiling Pt:377 °C (1013 hPa)
- Melting Pt:202 °C
- EINECS:209-488-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



