Order Entry
Export
ContactUsLinkComponent
Methyl-p-toluate
Methyl-p-toluate
Catalog # MOLEM29619711
Supplier:  Molekula
CAS Number:  
Methyl-p-toluate
Catalog # MOLEM29619711
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Methylbenzoic acid methyl ester, Methyl-4-methylbenzoate, p-Toluic acid methyl ester, 4-Toluic acid methyl ester, Methyl 4-methylbenzoate
 :
 :150.18
EINECS:202-784-1
 :GHS07
CAS Number:99-75-2
Formula:C₉H₁₀O₂
MDL Number:MFCD00008441
Boiling Pt:∼217 °C (1013 hPa)
Melting Pt:33…34 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:89