Methyl 2,4-dihydroxy-3,6-dimethylbenzoate
Catalog # MOLE73626632-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:196.2 g/mol
Density:1 g/cm³ (20 °C)
EINECS:225-193-0
CAS Number:4707-47-5
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00157202