
Benzyltrimethylammonium chloride
Catalog # MOLE10717982-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:185.7 g/mol
Melting Pt:238 °C
Density:1,07 g/cm³ (20 °C)
EINECS:200-300-3
:GHS07
Storage Temperature:Ambient
CAS Number:56-93-9
Formula:C₁₀H₁₆ClN
MDL Number:MFCD00011782