Order Entry
Export
Orders LinkContactUsLinkComponent
Cholic acid

Cholic acid

Glass bottle for solids, 100 g
Catalog # MOLE25751419-100G
Supplier:   Molekula
Cholic acid
Catalog # MOLE25751419-100G
Supplier:  Molekula
CAS Number:  81-25-4
Cholic acid

Glass bottle for solids, 100 g

CAS Number:  81-25-4
Price
220,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid
  • CAS Number:81-25-4
  • Formula:C₂₄H₄₀O₅
  • MDL Number:MFCD00003672
  • Molecular Weight:408.58 g/mol
  • Merck Index:13,02223
  • Boiling Pt:545 °C (1013 hPa)
  • Melting Pt:199 °C
  • Storage Temperature:Ambient
  • EINECS:201-337-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications