Indole-7-carbaldehyd
Glass bottle for solids, 1 g
Catalog # MOLE84400316-1G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 1H-Indole-7-carbaldehyde, Indole-7-carboxaldehyde
- CAS Number:1074-88-0
- Formula:C₉H₇NO
- MDL Number:MFCD01318152
- Molecular Weight:145.16 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



