2-Amino-4,6-dimethoxypyrimidine
Catalog # MOLEM16830266
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:155.16 g/mol
Boiling Pt:276 °C (1013 hPa)
Melting Pt:97 °C
Density:1,429 g/cm³ (–153 °C)
EINECS:252-969-6
CAS Number:36315-01-2
Formula:C₆H₉N₃O₂
MDL Number:MFCD00038832